Learn More
Hirschmann™ Class A USP Volumetric Flasks
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A USP Volumetric Flasks are manufactured from durable DURAN™ glass tubing and are available with a Batch Certificate of Performance, an Individual USP-Certificate of Performance, and a DAkkS-Certificate.
Brand:  Hirschmann™ 282019427
 View more versions of this product
Product Code. 10027652
  • 677.00 DKK / Each
Only null left
In Stock
Description
  • Blue graduation
  • With dated batch identification
  • With NS ground and polyethylene stopper
Specifications
Specifications
USP
2000 mL
370 mm
±0.5 mL
A
Polyethylene
Borosilicate Glass, Duran Glass
±0.5 mL
Yes
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hirschmann™ Class A USP Volumetric Flasks > Capacity: 2000mL