missing translation for 'onlineSavingsMsg'
Learn More

Glycine, ACS Reagent Grade, Scharlab

Product Code. 11374154
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Applications include analytical chemistry, in buffer solutions (for electrophoresis), for pharmaceutical use, and in the food industry.

  • ACS Reagent Grade
  • Synonyms: Aminoacetic acid, Glycocoll
  • Molecular Formula: C2H5NO2
  • Molar Mass: 75.07g/mol
  • CAS: 56-40-6
  • EIN ECS No.: 200-272-2
  • Substances Darkened by H2SO4: Passes test

Specifications

CAS 56-40-6
Melting Point 232°C to 236°C (decomposition)
pH 5.39 to 6.3 (5%, H2O)
Ignition Residue 0.05% max (600°C)
Molecular Formula C2H5NO2
MDL Number MFCD00008131
Quantity 1 kg
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
Solubility Information Soluble in water (225g/L at 20°C)
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILES NCC(O)=O
IUPAC Name 2-aminoacetic acid
Molecular Weight (g/mol) 75.07
PubChem CID 750
ChEBI CHEBI:15428
Formula Weight 75.07g/mol
Grade ACS Reagent
Concentration or Composition (by Analyte or Components) 99.7% min (assay, titration with HClO4)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycine, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.