Learn More
Glycine tert-butyl ester, 97%, Thermo Scientific Chemicals
Product Code. 11372957
Click to view available options
Quantity:
1 g
5 g
Glycine tert-butyl ester is used in the preparation of Schiff base by reacting with benzophenone. The resultant Schiff base undergoes asymmetric alkylation with arylmethyl bromides in the presence of O-allyl-N-(9-anthracenylmethyl)cinchonidinium bromide as chiral phase transfer catalyst to give guanidine-containing pentacyclic compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycine tert-butyl ester is used in the preparation of Schiff base by reacting with benzophenone. The resultant Schiff base undergoes asymmetric alkylation with arylmethyl bromides in the presence of O-allyl-N-(9-anthracenylmethyl)cinchonidinium bromide as chiral phase transfer catalyst to give guanidine-containing pentacyclic compound.
Solubility
Miscible with acetone and methanol. Slightly miscible with ehtanol. Immiscible with water.
Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6456-74-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00038194
InChI Key SJMDMGHPMLKLHQ-UHFFFAOYSA-N
Synonym tert-butyl glycinate, glycine tert-butyl ester, glycine, 1,1-dimethylethyl ester, tert-butyl aminoacetate, h-gly-otbu, t-butyl glycinate, glycin-tert-butyl ester, 1,1-dimethylethyl glycinate, glycine t-butyl ester, aminoacetic acid tert-butyl ester
PubChem CID 151417
IUPAC Name tert-butyl 2-aminoacetate
SMILES CC(C)(C)OC(=O)CN
Specifications
Boiling Point 29°C to 31°C (2mmHg)
Refractive Index 1.424
Beilstein 2039000
Solubility Information Miscible with acetone and methanol. Slightly miscible with ehtanol. Immiscible with water.
Density 0.96
Quantity 5 g
Formula Weight 131.18
Sensitivity Air sensitive
Percent Purity 97%
Chemical Name or Material Glycine tert-butyl ester