Learn More
GNE-477, MedChemExpress
MedChemExpress GNE-477 is a potent and efficacious dual PI3K (IC50=4 nM)/mTOR(Ki=21 nM) inhibitor.
Brand: Medchem Express HY-11042
This item is not returnable.
View return policy
Description
GNE-477 is a potent and efficacious dual PI3K (IC50=4 nM)/mTOR(Ki=21 nM) inhibitor.Specifications
GNE-477 | |
Yellow | |
C21H28N8O3S2 | |
DMSO : 16.67 mg/mL (33.03 mM; Need ultrasonic) | |
504.63 | |
98.7% | |
Solid |
1032754-81-6 | |
Research | |
50 mg | |
NC1=NC=C(C2=NC(N3CCOCC3)=C4C(C(C)=C(CN5CCN(S(=O)(C)=O)CC5)S4)=N2)C=N1 | |
504.63 | |
Research | |
Cancer-Kinase/protease |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
GNE-477, MedChemExpress