missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GnRHR Rabbit anti-Human, Clone: GNRHR/2982R, Novus Biologicals™
Rabbit Monoclonal Antibody
2107.94 DKK
Specifications
Antigen | GnRHR |
---|---|
Clone | GNRHR/2982R |
Concentration | 0.2 mg/ml |
Dilution | ELISA |
Applications | ELISA |
Description
GnRHR Monoclonal specifically detects GnRHR in Human samples. It is validated for ELISA.Specifications
GnRHR | |
0.2 mg/ml | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
GnRH receptor, GnRH-R, GNRHR1, gonadotropin-releasing hormone (type 1) receptor 1, gonadotropin-releasing hormone receptor, GRHR, leutinizing-releasing hormone receptor, LHRHRleutinizing hormone releasing horomone receptor, LRHR, luliberin receptor, type I GnRH receptor | |
Recombinant full-length human GnRHR protein | |
Primary | |
Store at 4°C. |
GNRHR/2982R | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
2798 | |
IgG | |
Protein A or G purified |