Learn More
Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A21125
This item is not returnable.
View return policy
Description
Goat anti-Mouse IgG1 Secondary Antibody, Alexa Fluor 594 conjugate, A-21125, from Invitrogen™. Species Reactivity: Mouse; Applications: Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Product Content Correction
Product Title
Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594, Invitrogen™ > 500 μg; Alexa Fluor 594
Spot an opportunity for improvement?Share a Content Correction