missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPAT2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-82903-0.1ML
This item is not returnable.
View return policy
Description
GPAT2 Polyclonal specifically detects GPAT2 in Human samples. It is validated for Western Blot.Specifications
GPAT2 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
150763 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
ALPAAB, BSCL, BSCL1, cancer/testis antigen 123, CT123, EC 2.3.1.15, glycerol-3-phosphate acyltransferase 2, mitochondrial, GPAT-2, xGPAT1 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human GPAT2 | |
0.1 mL | |
Primary | |
Human | |
Purified |