Learn More
Fisherbrand™ Grade 913 Lens Cleaning Tissue
Used for cleaning of optical lenses.
Brand:  Fisherbrand™ 0913F00034
Product Code. 10737643
  • 347.00 DKK / Pack of 25
Estimated Shipment: 18-06-2025
Explore more special offers
Add to basket
Description
Description
  • Very thin paper
  • Made with 100% Manilla fibers
  • Not fluffy
  • Weight: 12g/m2
Application
  • Cleaning of various optical lens
  • Cleaning of optical fibers filaments
Specifications
Specifications
Lens Cleaning Tissue
Cleaning of various optical lens (objective, microscopes binoculars, magnifying Glass, Glasses, etc.), cleaning of optical fibers filaments
Manilla Fibers
Documents
One moment while we fetch your results.
Product Title
Fisherbrand™ Grade 913 Lens Cleaning Tissue > Dimensions: 150L x 100mmW; Quantity: 625/Pk. Booklet