Learn More
Prestige Medical Green Gasket
Green Gasket for 121°C & 126°C models.
Brand: Prestige Medical 219776
This item is not returnable.
View return policy
Description
- Green Gasket for 121°C & 126°C models.
Specifications
Green Gasket | |
2 Pk. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Prestige Medical Green Gasket > Green Gasket; Quantity: 2 Pk.
Spot an opportunity for improvement?Share a Content Correction