Learn More
Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
Brand: Greiner Bio-One 450480
Description
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
- Tubes feature closure that acts as a valve, allowing appropriate capillary or funnel to be inserted without removing the cap
- Fit into a 13 Ă— 75mm carrier tube to facilitate bar code labeling, centrifugation, and storage
- Tubes with gel contain an inert barrier on the bottom that moves upward to form a stable barrier between the clot/cells and serum/plasma during centrifugation and transport
- All tubes require MiniCollect funnel or capillary collection device (sold separately)

Specifications
MiniCollect Capillary Blood Collection Tube | |
0.5 mL | |
Lavender |
K2 EDTA | |
Yes | |
Small volume blood collection |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.