Learn More
Deutsch Neumann™ Grey Natural Rubber Spacer
Brand:  Deutsch Neumann™ 2010011
Product Code. 10007990
  • 4.15 DKK / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Specifications
Gray
10Pack
18 mm
Natural Rubber
10/Pk.
Product Title
Deutsch Neumann™ Grey Natural Rubber Spacer > Height: 18mm; Quantity: 10/pk