Learn More
Gilac™ Grid
Grid is highly durable and shock resistant; it avoids any contact between food and exudates.
Brand:  Gilac™ G102521
 View more versions of this product
Product Code. 17958421
  • 16.75 DKK / Each
Only null left
Estimated Shipment: 18-08-2025
Description
  • Grid for flat trays.
  • Highly durable and shock resistant.
  • Enables the use of flat dishes as defrosting trays by separating the ice water from the product being defrosted.
Specifications
Specifications
Container Grid
Polyethylene
3 L Flat Tray
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Gilac™ Grid > For Use With: 3 L Flat Tray