Learn More
IKA™ Grinding Chambers
Brand: IKA™ 0002904100
5172.75 DKK valid until 2025-05-09
Use promo code "24143" to get your promotional price.
Description
Specifications
Grinding Chamber | |
Compatible with A11 basic analytical mill | |
Stainless Steel Inlet |
250 mL | |
Polycarbonate | |
A11.4 |