Learn More
MedChemExpress H-Gly-Gly-Pro-OH
MedChemExpress H-Gly-Gly-Pro-OH is a peptide with 3 amino acid.
Brand:  MedChemExpress HY-111922
Encompass_Preferred
Product Code. 16487257
  • POA / 5mg
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
97.0%
Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Peptides
Room temperature
14379-76-1
229.23 g/mol
5 mg
Glycyl-glycyl-L-proline
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.