missing translation for 'onlineSavingsMsg'
Learn More

H2DCFDA, >97%, Tocris Bioscience™

Product Code. 30002145
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Fluorescent ROS indicator; cell permeable

Fluorescent ROS indicator; becomes fluorescent when oxidized. Excitation wavelength ∽490 nm, emission ∽520 nm. Cell permeable.

Specifications

CAS 4091-99-0
Molecular Formula C24H16Cl2O7
Linear Formula C24H16Cl2O7
Quantity 100 mg
Synonym h2dcfda, 2-3,6-diacetoxy-2,7-dichloro-9h-xanthen-9-yl benzoic acid, 2',7'-dichlorohydrofluorescein diacetate, 2,7-dichlorodihydrofluorescein diacetate, benzoic acid, 2-3,6-bis acetyloxy-2,7-dichloro-9h-xanthen-9-yl, 2-3,6-bis acetyloxy-2,7-dichloro-9h-xanthen-9-yl benzoic acid, 2′,7′-dichlorofluorescin diacetate, 2′,7′-dichlorodihydrofluorescein diacetate, 2-3,6-diacetyloxy-2,7-dichloro-9h-xanthen-9-yl benzoic acid
Solubility Information Soluble to 100mM in DMSO and to 10mM in ethanol
InChI Key PXEZTIWVRVSYOK-UHFFFAOYSA-N
SMILES CC(=O)OC1=C(C=C2C(C3=CC(=C(C=C3OC2=C1)OC(=O)C)Cl)C4=CC=CC=C4C(=O)O)Cl
IUPAC Name 2-(3,6-diacetyloxy-2,7-dichloro-9H-xanthen-9-yl)benzoic acid
Molecular Weight (g/mol) 487.285
PubChem CID 77718
Formula Weight 487.29
Percent Purity 95%
Chemical Name or Material 2-[3,6-Bis(acetyloxy)-2,7-dichloro-9H-xanthen-9-yl]benzoic acid
Grade >97%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'