Learn More
Gilac™ HACCP Flat Tray with Lid and Grid
HACCP Flat Tray with Lid and Grid is a complete package with dimensions suitable for refrigerators.
145.00 DKK - 250.00 DKK
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | ||||
17948731
|
Gilac™
G190134 |
3 L | Red |
145.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17958841
|
Gilac™
G190228 |
5 L | Blue |
197.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17918891
|
Gilac™
G190328 |
8 L | Blue |
250.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Dimensions suitable for refrigerators.
- Can be used as a:
- defrosting tray thanks to its wide dividing grid that separates the ice water from the product being defrosted.
- preparation container for products that drip.
- ingredients tray.
- container for preserving comparison samples.
- fermenting and decanting container.
- Nesting and stacking with lid.
Specifications
HDPE | |
Dispensing Tray | |
Not Autoclavable |