missing translation for 'onlineSavingsMsg'
Learn More

Hairy (Drosophila) Antibody (1/24) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-76659-0.025ml

 View more versions of this product

Product Code. 18664819

  • 1495.00 DKK / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Hairy (Drosophila) Monoclonal specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hairy (Drosophila)
Monoclonal
1 mg/mL
Western Blot : 1:100 - 1:2000, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Mouse
Protein A purified
RUO
Primary
Drosophila
Purified
Western Blot, Immunohistochemistry, ChIP Assay
1/24
Unconjugated
PBS with 0.02% sodium azide
Hairy gene product
0.025 mL
Cellular Signaling, Developmental Biology, Epigenetics, Stem Cells
This antibody will stain Drosophila embryos.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hairy (Drosophila) Antibody (1/24) - BSA Free, Novus Biologicals™ > 0.025 mL; Hairy (Drosophila) Antibody; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.