Learn More
HAL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542755
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: EAAHRLLLEQ KVWEVAAPYI EKYRMEHIPE SRPLSPTAFS LQFLHKKSTK Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%
HAL Polyclonal antibody specifically detects HAL in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
HAL | |
Polyclonal | |
Unconjugated | |
HAL | |
hal; HIS; histidase; histidine ammonia lyase; histidine ammonia-lyase; histidinemia; Hsd; HSTD; Huth; zgc:136980 | |
Rabbit | |
Protein A | |
RUO | |
3034 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P42357 | |
HAL | |
Synthetic peptide directed towards the C-terminal of human HAL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |