Learn More
Macherey-Nagel™ Haloform Test Mixture in Methanol
Brand:  Macherey-Nagel™ 722371
Product Code. 10554542
  • 590.00 DKK / 1mL
Estimated Shipment: 22-05-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
For Headspace Analyses, for GC Environmental Analyses
Dichloromethane (795 μg/mL), Trichloromethane (75 μg/mL), 1,1,1-trichloroethane (67 μg/mL), Tetrachloromethane (80 μg/mL), Trichloroethene (73 μg/mL), Bromodichloromethane (100 μg/mL), Dibromochloromethane (122 μg/mL), Tetrachloroethene (81 μg/mL), Tribromomethane (145 μg/mL)
1 mL
Haloform Test Mixture in Methanol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Haloform Test Mixture in Methanol > Capacity: 1mL