Learn More
Hamilton™ 1700 Series Gastight™ Syringes
Ideal for dispensing both liquids and gases
Brand:  Hamilton™ CAL80900
 View more versions of this product
Product Code. 17894212
  • 2520.00 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Calibrated
  • Cemented Needle (N) and Luer Tip Cemented Needle (LTN)
  • 22s/22gauge w/point style-2
  • Gas Sterilizable
  • Precision-machined polymer plunger tip made of PTFE, which provides leak-free seal
  • Gastight fitting enable tip to wipe interior of syringe barrel free of sample
  • Reduces molecule deposition in syringe when samples have dissolved or suspended molecules
  • Volume range: 50μ to 500μL
Needles are 51mm unless user specified
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hamilton™ 1700 Series Gastight™ Syringes > Model: 1705; Cemented Needle (N); 1000 psig; 50Glc muL