Learn More
Hamilton™ Deep Well Plate
Brand:  Hamilton™ 6472-01
Product Code. 15167366
  • 995.00 DKK / Pack of 32
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Specifications
Polystyrene
Sterile
41 mm
Documents
One moment while we fetch your results.
Product Title
Hamilton™ Deep Well Plate > 1.2mL deep; Width: 127.5mm; Depth: 85.5mm; Height: 41.0mm