Learn More
Invitrogen™ Hand-Held Magnetic Plate Washer
Designed for use with the ProcartaPlex assay configured for magnetic separation. The handheld magnetic separator enables you to remove the liquid contents of the 96-well plate by simply decanting or flicking the contents into a sink and blotting off the remainder on a paper towel.
Brand: Invitrogen™ EPX-55555-000
7106.00 DKK valid until 2025-12-31
Use promo code "25044" to get your promotional price.
Description
- Reactivity: Canine, Human, Monkey, Mouse, Pig, Rat
- Please refer to the appropriate ProcartaPlex User Manual for details on running the assay
Specifications
Multiplex Immunoassay |