Learn More
Heathrow Scientific™ Hand Tally Counter
Count blood cells, bacterial colonies, and drops of liquid. Heathrow Scientific™ Hand Tally Counter can count any repetitive event with a quick push of a button.
Brand: Heathrow Scientific™ HS6594
Description
- Tallies up to 9999
- Features a quick-reset knob
- Lens for easy reading, and finger ring for a secure grip
- 4.6 dia. x 4.1cm W
Spécification
4 digits | |
Counts up to 9999 | |
4.1cm |
Manual click-counter | |
4.6cm | |
Finger ring, reset knob |
Suggestions de produits
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Days
59Hours
20Minutes
5Seconds
21
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.