missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANSELL™ Hazardous Chemicals AlphaTec™ Gloves
Description
- Offers superior protection against aliphatic, halogenated, and aromatic hydrocarbons (benzene, toluene, xylene), as well as concentrated mineral acids
- The glove is very flexible and provides excellent dexterity
- Designed for comfort, with a natural, curved ergonomic shape and soft feel, and offering easy donning and good grip
- The absence of latex proteins means there is no risk of latex allergies
Specifications
Specifications
Certifications/Compliance (Europe) | CE Number|EN 388:2016 +A1:2018|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|Category III|REACH Compliant |
Chlorinated | No |
Applications | Industrial |
Color | Black |
Latex Free | Yes |
Material | Viton |
Size | 10.5 |
EN388 Standard | 2120A |
EN ISO 374-1 | Type A |
EN ISO 374-1 CHEMICAL LETTER CODES | ABCDFLNS |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction