Learn More
asecos™ Hazardous Substances Rack
The Hazardous Substances Rack provides storage of flammable liquids and water pollutants.
Brand:  asecos™ 27868
 View more versions of this product
Product Code. 17816536
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • Rack with galvanised shelf as storage levels
  • Collecting sump
  • 100 mm ground clearance for transport with pallet truck
Specifications
Specifications
59.4 gal.
Steel
97.24 in.
97.24 in.
82.68 in.
32.13 in.
478.40 lb.
225 L
Drum Rack
2470.00 mm
2470.00 mm
2100.00 mm
816.00 mm
217 kg
Product Title
asecos™ Hazardous Substances Rack > Capacity: 225 L, 217 kg