Learn More
SymLine Safety Waste Caps
Safety Waste Caps safely connect the pipework to the waste container. The integrated exhaust filter blocks solvent vapours – for the protection of health and the environment.
620.00 DKK - 2945.00 DKK
Specifications
Type | Safety Waste Cap |
---|---|
For Use With (Application) | Waste Disposal System |
For Use With (Equipment) | SymLine System |
Product Code | Brand | Size | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Includes | Price | Quantity & Availability | |||||
16305988
![]() |
SymLine
106527 |
GL 45, GL 25 | Immersed filling lance, connector for exhaust filter, HDPE electrostatic conductive |
1260.00 DKK
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
17172566
![]() |
SymLine
306489 |
GL45 | 2 x PFA-fittings for capillaries with (OD) 1.6/2.3/3.2 mm |
620.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
17192566
![]() |
SymLine
306493 |
GL45 | 4 x PFA-fittings for capillaries with (OD) 1.6/2.3/3.2 mm |
705.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
17102576
![]() |
SymLine
306498 |
GL45 | 4 x PFA-fittings for capillaries with (OD) 1.6/2.3/3.2 mm, 1 x tube connector for tubes with (ID) 5.0 - 11.5 mm |
875.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
17182566
![]() |
SymLine
306492 |
GL45 | 7 x PFA-fittings for capillaries with (OD) 1.6/2.3/3.2 mm |
1225.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
10820411
![]() |
SymLine
106480 |
S60/61 | Electronic fill level control for connection table assembly |
2945.00 DKK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
Description
Specifications
Safety Waste Cap | |
Waste Disposal System | |
SymLine System |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
SymLine Safety Waste Caps