Learn More
Kartell™ Tank with Spigot
Designed to rationalize laboratory and small production line organization. Kartell™ Tank with Spigot gives the opportunity to optimize water, aqueous solutions and solvents availability in a reduced space.
Brand:  Kartell™ 0177100
 View more versions of this product
Product Code. 10640204
  • 1135.00 DKK / Pack of 4
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Description
  • Made of food grade High Density PE
  • Includes comfortable handle for displacement
  • The spigot is placed in a position allowing the tank to stand, as to facilitate the use of these tanks in packaging and transporting liquids
  • Suitable for foodstuff
Specifications
Specifications
HDPE
470 mm
180 mm
20 L
370 mm
4
Product Title
Kartell™ Tank with Spigot > Capacity: 20L