missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE, Carboy

Capacity Metric: 10 L

Brand:  Kautex™ 2000084133

Product Code. 10291501

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Natural color
  • Blue printed graduation markings
  • Screwed-on closure
  • Available with stop cock that comes with an yellow dust cap (dust cap does not seal the carboy)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 L
206 mm
6/Cs.
Carboy
50 mm
HDPE
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers