Specifications
250 mL | |
150W |
Medline Heating Stirrer Mantle (250ml)
File size: 214.1 kB
Product Content Correction
Product Title
Medline™ Heating Mantles > Capacity: 250mL; Wattage: 150W
Spot an opportunity for improvement?Share a Content Correction