Learn More
Heidolph™ Hei-VAP Basic Package
Hei-VAP Core with hand lift and vertical glassware set G3.
Brand: Heidolph™ 571-34300-00
This item is not returnable.
View return policy
Description
- Hei-VAP Core with hand lift and vertical glassware set G3
- Vacuum pump Rotavac Valve Tec
- Manual vacuum controller
- Tubing set
Specifications
EU Plug | |
Hand Lift Rotary Evaporator |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
- Multiple Manufacturers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Heidolph™ Hei-VAP Basic Package > EU Plug
Spot an opportunity for improvement?Share a Content Correction