Learn More
Heidolph™ Hei-VAP Core HL G3B XL
An operating concept that is focused on the essentials: clear and with access to the basic parameters rotation speed and heating bath temperature at any time.
Brand:  Heidolph™ 571-01315-00
29630.58 DKK valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 15978882
  • 34955.00 DKK / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Description
  • Clear digital display showing actual and set values
  • Two separate knobs with dynamic control for setting the rotation speed and heating bath temperature: fast turning for large jumps, slow turning for precise setting of the set values
  • Accidental changing of the values can be prevented by the lock function
  • Intuitive LED ring light system makes activated functions clearly visible even from a distance
  • Easy adjustment of the immersion depth up to 155 mm and the inclination angle from 20° to 80°
  • Standby function and residual heat warning at heating bath temperatures above 50°C
  • Hand lift model with transparent plastic coated G3 XL vertical glassware - achieve higher evaporation rates due to expanded cooling surface of 2,200 cm2
  • The standby button stops all functions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EU Plug
Hand Lift Rotary Evaporator
Product Title
Heidolph™ Hei-VAP Core HL G3B XL > EU Plug