Learn More
Heidolph™ Instruments Hei-VAP Reaction Extraction-Chill Package, EU-plug
The Hei-VAP Reaction Extraction-Chill Package features a motor lift, chiller, and vapor temperature sensor.
Brand:  Heidolph™ Instruments 574-23600-00
Product Code. 17373330
  • 112565.00 DKK / Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Hei-VAP Expert Control with motor lift and vertical glassware set G6
  • Vacuum pump Rotavac Vario Tec
  • Vapor temperature sensor
  • Chiller Hei-CHILL 350
  • RS 232 cable
  • Thermal fluid Kyro 30 (10 L)
  • Tubing set
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Motor Lift Rotary Evaporator
EU-plug
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Heidolph™ Instruments Hei-VAP Reaction Extraction-Chill Package, EU-plug > Type: Motor Lift Rotary Evaporator