Learn More
Heidolph™ Instruments Hei-VAP Ultimate Control HL G1B, EU-plug
The Hei-VAP Ultimate Control HL G1B adds numerous smart features to the Hei-VAP Ultimate.
Brand:  Heidolph™ Instruments 575-02110-00
Product Code. 17333350
  • 46970.00 DKK / Each
Estimated Shipment: 10-06-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Adds numerous smart features to the Hei-VAP Ultimate, as well as an overview of all parameters and the current process.
  • 7 in. touch display for controlling the entire system including the vacuum and the cooling temperature
  • Hand lift model with transparent plastic coated G1 diagonal glassware
  • For precise monitoring, the pictogram on the display also symbolizes active functions
  • Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
  • Setting of values either via the touch display or the separate quick access knobs
  • Direct access to two favorites on the start screen
  • Expandable solvent library. You can also transfer your own favorites to other devices using a USB stick
  • Define and save own evaporation processes (ramps)
  • Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation
  • USB and Micro SD plus LAN / RS 232 interfaces (Lab 4.0 ready)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 L
Hand Lift Rotary Evaporator
EU-plug
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Heidolph™ Instruments Hei-VAP Ultimate Control HL G1B, EU-plug > Type: Hand Lift Rotary Evaporator; Capacity (Metric): 5 L