missing translation for 'onlineSavingsMsg'
Learn More
HEPES sodium salt, >98%, MP Biomedicals™ Product Code.: 11481952

HEPES sodium salt, >98%, MP Biomedicals™

Product Code. 11481952 Shop All MP Biomedical Products
250 g
Click to view available options
Quantity:
100 g
250 g
This item is not returnable. View return policy

Product Code. 11481952

Brand: MP Biomedicals 0210559383

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium saltShow MoreShow Less
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range >98%
Formula Weight 260.3
Chemical Name or Material HEPES sodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HEPES sodium salt, >98%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.