Learn More
Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations.
Brand:  Thermo Scientific™ 50140095
 View more versions of this product
Product Code. 15188912
  • 9527.65 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations. Options include media valves, solenoid valve, stainless-steel work surface wraps, side window modifications, UV and UV night covers, windscreen protection kits and additional accessories.
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Heraguard ECO 1.8
Retrofit Kit UV-Radiator
UV Options
Field Installed