Learn More
HICAL Acids Mix, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep HICAL-ACIDS-5
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
2,4,5-Trichlorophenol | 95-95-4 | 2000μg/mL |
2,4,6-Trichlorophenol | 88-06-2 | 2000μg/mL |
2,4-Dichlorophenol | 120-83-2 | 2000μg/mL |
2,4-Dimethylphenol | 105-67-9 | 2000μg/mL |
2,4-Dinitrophenol | 51-28-5 | 2000μg/mL |
2-Chlorophenol | 95-57-8 | 2000μg/mL |
2-Methylphenol | 95-48-7 | 2000μg/mL |
2-Nitrophenol | 88-75-5 | 2000μg/mL |
4,6-Dinitro-2-methylphenol | 534-52-1 | 2000μg/mL |
4-Chloro-3-methylphenol | 59-50-7 | 2000μg/mL |
4-Methylphenol | 106-44-5 | 2000μg/mL |
4-Nitrophenol | 100-02-7 | 2000μg/mL |
Benzoic acid | 65-85-0 | 2000μg/mL |
Pentachlorophenol | 87-86-5 | 2000μg/mL |
Phenol | 108-95-2 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
HICAL Acids Mix | |
2000μg/mL | |
MeCl | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
95-95-4, 88-06-2, 120-83-2, 105-67-9, 51-28-5, 95-57-8 | |
5mL amber prescored glass ampule | |
5 mL |
Safety and Handling
- HICAL ACIDS MIX
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Skin sensitiser Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : dichloromethane, 2,4-dichlorophenol, 2,4,6-trichlorophenol,2-chlorophenol, DNOC
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product intended for laboratory use only