Få mere at vide
Thermo Scientific™ High Performance Microtube with Snap-on Seal Closure Artikelnummer.: 15361379
Thermo Scientific™ High Performance Microtube with Snap-on Seal Closure
Artikelnummer. 15361379 Shop alle Thermo Scientific produkter
S110-AT, S100-AT4, S100-AT5, S100-AT6, and S80-AT3 rotors with adapters. S55-A2 and S45-A rotor are used without adapters.
:
S110-AT, S100-AT4, S100-AT5, S100-AT6, and S80-AT3 rotors with adapters. S55-A2 and S45-A rotor are used without adapters.
Artikelnummer. 15361379
Brand: Thermo Scientific™ 314352H01
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Recommended when using microcentrifuge tubes in Thermo Scientific micro-ultracentrifuge rotors.
  • 1.5 mL High Performance Microtube with Snap-on Seal Closure.
  • Recommended when using microcentrifuge tubes in Thermo Scientific micro-ultracentrifuge rotors.
  • The maximum RCF of this microtube is 201,000 x g
TRUSTED_SUSTAINABILITY
Tekniske data
Type Mikrocentrifugerør
Til brug med (udstyr) S110-AT, S100-AT4, S100-AT5, S100-AT6, and S80-AT3 Rotors with Adapters. S55-A2 and S45-A Rotor Are Used Without Adapters.
Volumen 1.5 mL
Maks. RCF 201,000 x g
Maks. Hastighed 55,000 rpm
Diameter (metrisk) Udvendig 11 mm
Længde (metrisk) 42 mm
nr. pr. pakke 100
Korrektion af produktindhold
Produkttitel
Thermo Scientific™ High Performance Microtube with Snap-on Seal Closure >