Learn More
Masterflex™ High Pressure Tubing with Luer Ends
Use to connect to bio-bags or a sampling tube. Fitting is molded onto the tubing for a seamless transition
Brand: Masterflex™ 30526-14
This item is not returnable.
View return policy
Description
- Plastic Luer Assemblies
- Get a seamless transition from source to system
- Each assembly comes packaged individually; can be sterilized
- High pressure tubing has luer fittings at each end
- Male luer lock x female luer in varying lengths from 10 to 48 in.
- Sterilize by ETO
Specifications
PVC | |
High Pressure tubing with Luer Ends | |
2 ports: 1 Male, 1 female luer lock end | |
900 psi |