Learn More
Nabertherm™ High-Temperature Furnaces with Molybdenum Disilicide Heating Elements and Fiber Insulation
Due to their solid construction and compact stand-alone design, these high-temperature furnaces are perfect for processes in the laboratory where the highest precision is needed. Oustanding temperature uniformity and practical details set very high quality benchmarks.
180935.00 DKK - 267975.00 DKK
Specifications
Includes | Molybdenum Disilicide Heating Elements and Fiber Insulation |
---|---|
Voltage | 400 V |
Product Code | Brand | Capacity (Metric) | Temperature (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Temperature (Metric) | Price | Quantity & Availability | |||||
17347839
|
Nabertherm™
HT0246VNA |
8 L | 1600°C |
180935.00 DKK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
17357839
|
Nabertherm™
HT0247VNA |
8 L | 1750°C |
187130.00 DKK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
17367839
|
Nabertherm™
HT0248VNA |
8 L | 1800°C |
244965.00 DKK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
17377839
|
Nabertherm™
HT0356VNA |
16 L | 1600°C |
199370.00 DKK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
17387839
|
Nabertherm™
HT0357VNA |
16 L | 1750°C |
208015.00 DKK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
17397839
|
Nabertherm™
HT0358VNA |
16 L | 1800°C |
267975.00 DKK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
Description
- Dual shell housing with fan cooling for low shell temperatures
- Heating from both sides via molybdenum disilicide heating elements
- High-quality fiber insulation backed by special insulation
- Side insulation constructed with tongue and groove blocks provides for low heat loss to the outside
- Long-life roof insulation with special suspension
- Chain-guided parallel swivel door for defined opening and closing of the door
- Vapor vent in the furnace roof with motorized exhaust air flaps, controlled via the extra function of the controller
- Heating elements switched via thyristors
Specifications
Molybdenum Disilicide Heating Elements and Fiber Insulation | |
400 V |