missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3.3 Rabbit anti-Human, Mouse, Rat, Clone: 5C5B8, Novus Biologicals™
Rabbit Monoclonal Antibody
1460.01 DKK - 3692.13 DKK
Specifications
Antigen | Histone H3.3 |
---|---|
Clone | 5C5B8 |
Dilution | Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18360423
|
Bio-Techne
NBP3-16489-20UL |
20 μg |
1460.01 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18310556
|
Bio-Techne
NBP3-16489-100UL |
100 μg |
3692.13 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H3.3 Monoclonal antibody specifically detects Histone H3.3 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Histone H3.3 | |
Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
H3.3A, H3F3, H3F3A, Histone H3.3, Histone H3.3A, Histone H3.3B, MGC87782, MGC87783 | |
A synthetic peptide corresponding to a sequence within amino acids 57-136 of human Histone H3.3 (P84243). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
5C5B8 | |
ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Chromatin Research, Epigenetics | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3020 | |
IgG | |
Affinity purified |