Learn More
Novus Biologicals™ HMGCL Overexpression Lysate
Brand: Novus Biologicals™ NBL1-11617
This item is not returnable.
View return policy
Description
The HMGCL Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC23817 and based on accession number NM_191. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
Western Blot | |
RIPA buffer | |
Homo sapiens 3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase (HMGCL), mRNA. | |
Cancer, Signal Transduction |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction