Learn More
Hoefer™ Blotting Paper
Cellulose papers precut for convenience
Brand:  Hoefer™ TE76
 View more versions of this product
Product Code. 10762097
  • 670.00 DKK / Pack of 25
In Stock
Explore more special offers
Description
  • For Western Blotting
TRUSTED_SUSTAINABILITY
Specifications
Specifications
21 cm
TE77 Semi-Dry Blotting Units
Cellulose
25 cm
Sheet
Documents
One moment while we fetch your results.
Product Title
Hoefer™ Blotting Paper > 21 x 26cm