Learn More
Hoefer™ Replacement Parts for HE 99X Max Submarine Electrophoresis Unit
Various replacement parts for Submarine Electrophoresis Unit
2230.00 DKK - 2905.00 DKK
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
11569563
|
Hoefer™
HE96X |
Lid Assembly | Lid, red and black high voltage Leads and two lid plugs |
2230.00 DKK
Each |
Estimated Shipment: 21-03-2025
Log in to see stock. |
||||
11579543
|
Hoefer™
HE90X |
Buffer Chamber Assembly | 4 Leveling feet, 2 banana plugs with Nickel and Nylon washers and 2 Platinum wire electrode strip assemblies |
2905.00 DKK
Each |
Estimated Shipment: 21-03-2025
Log in to see stock. |
||||
Description
Replacement Parts
- Buffer chamber assembly (Includes 4 leveling feet, 2 banana plugs with nickel and nylon washers, and 2 platinum wire electrode strip assemblies)
- Lid assembly (Includes lid, red and black high-voltage leads, and two lid plugs)
Specifications
HE99X Max Submarine Electrophoresis Unit |