Learn More
Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates
Use with the SE 250 vertical electrophoresis unit. Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates come in a variety of styles for particular needs.
Brand:  Hoefer™ SE202N-10
 View more versions of this product
Product Code. 10585585
  • 1495.00 DKK / Pack of 10
Only null left
Estimated Shipment: 25-08-2025
Description
  • 10 x 8 cm
Specifications
Specifications
SE 250 Vertical Electrophoresis Unit
Notched Alumina Plate
Alumina
10/Pk.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates > Alumina plate; Notched; 10 x 8cm; 10/Pk.