Learn More
Cytiva SE 400 Vertical Unit
The sturdier vertical slab gel unit was designed with durability and cost in mind. Injection-moulded of tough, impact-resistant plastics, it will withstand the rigours of daily lab use and teaching labs for years
Brand:  Cytiva SE400-15-1.5
Product Code. 12944840
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • SE 400 Vertical Units run one 16-cm gel or two gels with an accessory divider plate.
  • Its low cost makes the SE 400 appropriate for labs requiring multiple units when temperature control is not crucial.
  • The unit base is also a convenient casting stand for single gels. To cast multiple gels for one or more units, use the multiple gel casters.
  • Buffer volume: 350 ml (Minimum upper buffer volume) 150 ml (Minimum lower buffer volume) 350 ml (Maximum lower buffer volume)
  • Total Separation Time: 3-5 h