Learn More
Sperian™ Honeywell™ FFP3 ML Respirator with Valve
Click to view available options
:
Filter Type: FPP3
Description
- Rigid shell - maintains shape even in moist and heavy loading environments
- Ultra-soft hypoallergenic Willtech™ flange seal with absorption properties and a tight sealing film for exceptional comfort and perfect fit
- Preformed and colored nose bridge to save time adjusting the mask and instantly identify its protection level
- Large internal volume to easily communicate and breathe
- Shielded exhalation valve with an exclusive design for improved protection
- Adjustable braids to easily and efficiently fit the mask
- No latex, no silicone, no PVC
Specifications
Specifications
Aerosol Type | Solid and/or Liquid |
Certifications/Compliance | EN 149:2001A1:2009 |
Disposable | Single-use |
Product Type | Respiratory Protection Molded Mask |
For Use With (Application) | Protects against dusts, mists and fumes containing calcium carbonate, clay, kaolin, cellulose, cotton, flour, ferrous Metals, cement, sodium silicate, sulphur, Glass fibers and plastics, hardwood, coal, quartz, non ferrous Metals, copper, aluminum, barium, titanium, vanadium, chrome, manganese, molybdenum, antimony, nickel, platinum, rhodium, strychnine, uranium, moderately radio-active particles, vegetal and mineral oils, Metal-working fluids (this list is non-exhaustive) |
Quantity | 5/Pk. |
Weight (Metric) | 0.168 kg |
Product Content Correction
Product Title
Sperian™ Honeywell™ FFP3 ML Respirator with Valve >
Spot an opportunity for improvement?Share a Content Correction