Learn More
Medicom™ Hopen™ Pocketless Hygiene Gown
Medicom™ Hopen™ Pocketless Hygiene Gown available in three distinct colors
Brand:  Medicom™ 40.301/06
 View more versions of this product
Product Code. 16564051
  • 379.00 DKK / Pack of 50
Only null left
Estimated Shipment: 30-09-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • The Pocketless Hygiene gown protects the working environment from particles coming from the body of the operator
  • Allows the user to be protected from the risks of soiling related to the activity being carried out
  • Pleasant and light, polypropylene fabric is extremely breathable
  • Collared model without flap and elasticated at the wrists
Specifications
Specifications
3X-Large
Category 1 Personal Protective Equipment
None
Hygiene Gown
White
Single-use
Elastic
Manufactured under an ISO 9001 Certified Quality System
Made in China
Unisex
Individual, Bag
Polypropylene
Press Studs
Non-sterile
Sewn
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Medicom™ Hopen™ Pocketless Hygiene Gown > White, 3X-Large