missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC11 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1485.92 DKK - 3498.83 DKK
Specifications
Antigen | HOXC11 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18310774
|
Bio-Techne
NBP3-04552-20UL |
20 μL |
1485.92 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18323205
|
Bio-Techne
NBP3-04552-100UL |
100 μL |
3498.83 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXC11 Polyclonal specifically detects HOXC11 in Human samples. It is validated for Western Blot.Specifications
HOXC11 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
3227 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
homeo box C11, homeobox C11, Homeobox protein Hox-3H, homeobox protein Hox-C11, HOX3H, MGC4906 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human HOXC11 (NP_055027.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |