Learn More
Thermo Scientific HPLC COLUMN HYPERREZ XP PARTICLEsize 8µm 250mm x 4.6mm I.D.
Brand:  Thermo Scientific 68008-254680
12074.97 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
HPLC COLUMN HYPERREZ XP PARTICLEsize 8µm 250mm x 4.6mm I.D.
Product Code. 11587541
  • 12135.34 DKK / Each
Estimated Shipment: 16-06-2025
Explore more special offers
Product Title
Thermo Scientific HPLC COLUMN HYPERREZ XP PARTICLEsize 8µm 250mm x 4.6mm I.D. >