missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSCB Mouse anti-Human, Clone: OTI2D10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF507276
This item is not returnable.
View return policy
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May act as a co-chaperone in iron-sulfur cluster assembly in mitochondria.Specifications
HSCB | |
Monoclonal | |
Unconjugated | |
HSCB | |
DNAJC20, HSC20, JAC1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
150274 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2D10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8IWL3 | |
HSCB | |
Full length human recombit protein of human HSCB produced in HEK293T cell | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |